Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

668265

Sigma-Aldrich

N-Boc-2-piperidineethanol

97%

Synonym(s):

1-tert-Butoxycarbonyl-2-piperidineethanol, 2-(2-Hydroxyethyl)piperidine-1-carboxylic acid tert-butyl ester, N-Boc-2-(2-hydroxymethyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23NO3
CAS Number:
Molecular Weight:
229.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.472

bp

100-110 °C/0.3 mmHg

density

1.032 g/mL at 25 °C

functional group

hydroxyl

SMILES string

CC(C)(C)OC(=O)N1CCCCC1CCO

InChI

1S/C12H23NO3/c1-12(2,3)16-11(15)13-8-5-4-6-10(13)7-9-14/h10,14H,4-9H2,1-3H3

InChI key

LTVQOFUGXMVESU-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Vasopressin1b receptor antagonists
  • CXCR4 antagonists as anti-HIV agents
  • Coumarin-based inhibitors of inducible nitric oxide synthase
  • Selective thrombin inhibitors

Reactant for:
  • N-directed hydroboration for synthesis of amino alcohols
  • Enantioselective synthesis of sedamine and allosedamine

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

668265-VAR:
668265-BULK:
668265-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service