Skip to Content
Merck
All Photos(3)

Key Documents

Safety Information

666017

Sigma-Aldrich

Potassium cyclopentyltrifluoroborate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9BF3K
Molecular Weight:
176.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

>300 °C

SMILES string

[K+].F[B-](F)(F)C1CCCC1

InChI

1S/C5H9BF3.K/c7-6(8,9)5-3-1-2-4-5;/h5H,1-4H2;/q-1;+1

InChI key

MUVSOIJDVXEYST-UHFFFAOYSA-N

Application

Organotrifluoroborate involved in:
  • Direct alkylation of heteroaryls
  • Oxidation via Oxone or via copper(II) salts and Dess-Martin periodinane
  • Cross-coupling with aryl chlorides
  • Suzuki cross-coupling

Organotrifluoroborates as versatile and stable boronic acid surrogates.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

666017-BULK:
666017-VAR:
666017-5G:
666017-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service