Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

665320

Sigma-Aldrich

4-Bromo-2-methylbenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(CH3)CN
CAS Number:
Molecular Weight:
196.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

65-69 °C

functional group

bromo
nitrile

SMILES string

Cc1cc(Br)ccc1C#N

InChI

1S/C8H6BrN/c1-6-4-8(9)3-2-7(6)5-10/h2-4H,1H3

InChI key

LPEBMDFRIKYFCF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

665320-BULK:
665320-25G:4548173228440
665320-VAR:
665320-5G:4548173228433


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service