Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

662577

Sigma-Aldrich

4-Fluoro-2-trifluoromethyl benzo-6-d1-nitrile-13C, 15N

99 atom % 13C, 98 atom % D, 98 atom % 15N, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC7DH2F415N
CAS Number:
Molecular Weight:
192.10
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C
98 atom % 15N
98 atom % D

Assay

97% (CP)

form

solid

mp

44-45 °C

mass shift

M+3

SMILES string

[2H]c1cc(F)cc(c1[13C]#[15N])C(F)(F)F

InChI

1S/C8H3F4N/c9-6-2-1-5(4-13)7(3-6)8(10,11)12/h1-3H/i1D,4+1,13+1

InChI key

LCCPQUYXMFXCAC-QCUBOOBBSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

662577-VAR:
662577-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service