662550
Lumidot™ CdSe-6, quantum dot nanoparticles kit
core-type, fluorescence λem 480-640 nm
Synonym(s):
QD, Quantum dot semiconductor nanocrystal
About This Item
Recommended Products
form
dispersion
shelf life
1 yr
concentration
5 mg/mL in toluene
bp
110 °C
density
0.865 g/mL at 25 °C
fluorescence
FWHM <40 nm
λem 480-640 nm
storage temp.
2-8°C
General description
Kit contains six toluene solutions of CdSe quantum dot nanocrystals with fluorescence emission maxima spanning visible spectrum:
- Lumidot™ CdSe 480 5 ml, 5 mg/ml, 480 nm
- Lumidot™ CdSe 520 5 ml, 5 mg/ml, 520 nm
- Lumidot™ CdSe 560 5 ml, 5 mg/ml, 560 nm
- Lumidot™ CdSe 590 5 ml, 5 mg/ml, 590 nm
- Lumidot™ CdSe 610 5 ml, 5 mg/ml, 610 nm
- Lumidot™ CdSe 640 5 ml, 5 mg/ml, 640 nm
Lumidot™ CdSe 640 nanocrystals are surface-stabilized with hexadecylamine/trioctylphosphine oxide ligands.
Typical nanocrystal sizes range from 2.5 nm (Lumidot™ CdSe 480) to 6.5 nm (Lumidot™ CdSe 640). The soft ligands increase hydrodynamic diameter of the nanocrystals by ~4.5-5 nm.
Features and Benefits
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
Flash Point(F)
39.2 °F
Flash Point(C)
4 °C
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PDSCL
Poisonous substance
FSL
Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid
JAN Code
662550-BULK:
662550-VAR:
662550-1EA:
662550-1KT:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service