Skip to Content
Merck
All Photos(1)

Documents

Safety Information

662127

Sigma-Aldrich

Palmitoyl-1,2,3,4-13C4-L-carnitine hydrochloride

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C19H45NO4 · HCl
Molecular Weight:
440.04
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

mp

166 °C

mass shift

M+4

SMILES string

Cl.CCCCCCCCCCCC[13CH2][13CH2][13CH2][13C](=O)O[C@H](CC([O-])=O)C[N+](C)(C)C

InChI

1S/C23H45NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(27)28-21(19-22(25)26)20-24(2,3)4;/h21H,5-20H2,1-4H3;1H/t21-;/m1./s1/i16+1,17+1,18+1,23+1;

InChI key

GAMKNLFIHBMGQT-GDTWUBLWSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

662127-BULK:
662127-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service