Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

655236

Sigma-Aldrich

(S)-(+)-2-Hexanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1718997
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D +12°, neat

refractive index

n20/D 1.415 (lit.)

bp

137-138 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCC[C@H](C)O

InChI

1S/C6H14O/c1-3-4-5-6(2)7/h6-7H,3-5H2,1-2H3/t6-/m0/s1

InChI key

QNVRIHYSUZMSGM-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

655236-VAR:
655236-BULK:
655236-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service