Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

654299

Sigma-Aldrich

(R)-(+)-o-Tolyl-CBS-oxazaborolidine solution

0.5 M in toluene

Synonym(s):

(R)-(+)-3,3-Diphenyl-1-o-tolyl-tetrahydropyrrolo(1,2-c)(1,3,2)oxazaborole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24BNO
CAS Number:
Molecular Weight:
353.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in toluene

refractive index

n20/D 1.514

density

0.900 g/mL at 25 °C

functional group

phenyl

SMILES string

[H][C@]12CCCN1B(OC2(c3ccccc3)c4ccccc4)c5ccccc5C

InChI

1S/C24H24BNO/c1-19-11-8-9-16-22(19)25-26-18-10-17-23(26)24(27-25,20-12-4-2-5-13-20)21-14-6-3-7-15-21/h2-9,11-16,23H,10,17-18H2,1H3/t23-/m1/s1

InChI key

XHMKFCAQQGBIPZ-HSZRJFAPSA-N

Application

When protonated with trifluoromethanesulfonimide, chiral oxazaborolidines generate chiral Lewis acids, which have demonstrated great utility in the enantioselective Diels-Alder reaction.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

FSL

Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

654299-25ML:4548173219875
654299-5ML:4548173219882
654299-VAR:
654299-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Do Hyun Ryu et al.
Journal of the American Chemical Society, 125(21), 6388-6390 (2003-06-06)
The strong acid triflimide ((CF3SO2)2NH) protonates chiral oxazaborolidines to form superactive, stable, chiral Lewis acids which are highly effective catalysts for a wide variety of enantioselective Diels-Alder reactions, documented herein by more than 20 examples.
Santanu Mukherjee et al.
Organic letters, 12(3), 632-635 (2010-01-13)
Diels-Alder reactions of various combinations of maleimides and 1,3-dienes with cationic oxazaborolidines as catalysts have been shown to be highly efficient and enantioselective.
Stemmler, R.T.
Synlett, 997-997 (2007)

Articles

we are pleased to offer both enatiomers of 2-methyl-CBS-oxazaborolidine as a dry reagent, in addition to our current offerings as a 1M solution in toluene.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service