Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

649791

Sigma-Aldrich

Poly(3,4-ethylenedioxythiophene)-block-poly(ethylene glycol) solution

1 wt % dispersion in nitromethane, contains p-toluenesulfonate as dopant

Synonym(s):

Aedotron P-NM, PEDOT-block-PEG, PEDOT-b-PEG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103

contains

p-toluenesulfonate as dopant

concentration

1 wt % dispersion in nitromethane

work function

4.19 eV

conductivity

10-4-10-3 S/cm

density

1.127 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Please follow this link for Aedotron/Oligotron selection guide table.

Application

Nitromethane dispersions can be used to cast thin films on glass, ITO, silicon wafers and many polymeric substrates. Compatibility of the polymeric substrate with nitromethane should be tested in each case. Coat the substrate with a "pool" of the nitromethane dispersion. Thin transparent films are typically spun at 1000 RPM or higher speeds, but more conducting films can be obtained at lower speeds. A single layer spun at 1000 RPM is typically 75 nm thick.
Conducting polymer. Useful as anti-static layers, hole-transport layers in OLED′s and various organic electronics applications (flexible circuits, capacitors, batteries).

Caution

Store this product at room temperature (do not refrigerate or freeze). Some settling will normally occur. Agitate, sonicate, and filter prior to use. A plug of glass wool in a pipette can be used for easy filtration. The shelf life of this product will vary depending on how it is stored and handled. Product should be discarded if heavy aggregation occurs, or if sonication / filtering are inadequate to produce a good casting solution.

Legal Information

Aedotron is a trademark of TDA Research, Inc.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

649791-BULK:
649791-VAR:
649791-25G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service