Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

644927

Sigma-Aldrich

3-Methylpyrazole-5-carboxylic acid

97%

Synonym(s):

5-Methylpyrazole-3-carboxylic acid, AS057278, MPC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

236-240 °C (lit.)

functional group

carboxylic acid

SMILES string

Cc1cc([nH]n1)C(O)=O

InChI

1S/C5H6N2O2/c1-3-2-4(5(8)9)7-6-3/h2H,1H3,(H,6,7)(H,8,9)

InChI key

WSMQKESQZFQMFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

644927-5G:
644927-VAR:
644927-BULK:
644927-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiziana Adage et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 18(3), 200-214 (2007-08-08)
Non-competitive N-methyl-d-aspartate (NMDA) blockers induce schizophrenic-like behavior in healthy volunteers and exacerbate symptomatology in schizophrenic patients. Hence, a compound able to enhance NMDA neurotransmission by increasing levels of d-serine, an endogenous full agonist at the glycine site of the NMDA
A McAllister et al.
Biochemical pharmacology, 31(1), 63-68 (1982-01-01)
The administration of the antilipolytic agents sodium nicotinate (1 mmole/kg i.p.) or sodium 5-methylpyrazole-3-carboxylate (0.5 or 1.0 mmole/kg i.p.) to alloxan-diabetic rats produced a significant reduction in the plasma concentration of free fatty acids and a slight reduction in blood
Plasma prostaglandin levels in rats with diabetes mellitus and diabetic ketoacidosis.
L Axelrod et al.
Diabetes, 31(11), 994-1001 (1982-11-01)
Takeshi Fukushima et al.
Chirality, 21(4), 468-472 (2008-07-26)
L-Kynurenine (KYN), a tryptophan metabolite, is metabolized to kynurenic acid (KYNA), which is an antagonist of N-methyl-D-aspartate and alpha7 nicotinic acetylcholine receptors, by kynurenine aminotransferase (KAT) I and KAT II. In this study, optically pure KYN, namely L-KYN or D-KYN
Michael Williams
Biochemical pharmacology, 78(11), 1360-1365 (2009-07-14)
Antipsychotics, the drugs used currently for the treatment of schizophrenia, produce their therapeutic effects via the blockade of dopamine receptors. These compounds are, however, limited in their therapeutic efficacy and have side effect liabilities that also limit their use. Agents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service