Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

640107

Sigma-Aldrich

5-Amino-2-(trifluoromethyl)pyridine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5F3N2
CAS Number:
Molecular Weight:
162.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

39-43 °C (lit.)

functional group

fluoro

SMILES string

Nc1ccc(nc1)C(F)(F)F

InChI

1S/C6H5F3N2/c7-6(8,9)5-2-1-4(10)3-11-5/h1-3H,10H2

InChI key

IBOZOWZSXZNIHI-UHFFFAOYSA-N

Application

Employed in a covenient, one-pot, synthesis of azaindoles.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

640107-5G:
640107-VAR:
640107-1G:
640107-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 46, 2283-2283 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service