Skip to Content
Merck
All Photos(3)

Documents

Safety Information

640085

Sigma-Aldrich

6-(Trifluoromethyl)pyridine-3-carboxaldehyde

95%

Synonym(s):

6-(Trifluoromethyl)nicotinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4F3NO
CAS Number:
Molecular Weight:
175.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

52-56 °C

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc(nc1)C(F)(F)F

InChI

1S/C7H4F3NO/c8-7(9,10)6-2-1-5(4-12)3-11-6/h1-4H

InChI key

MRPAGRCGPAXOGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

199.9 °F - closed cup

Flash Point(C)

93.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

640085-5G:
640085-1G:
640085-VAR:
640085-BULK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service