Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

637971

Sigma-Aldrich

trans-2-(3-Fluorophenyl)vinylboronic acid

≥95%

Synonym(s):

(E)-2-(3-Fluorophenyl)ethenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8FBO2
CAS Number:
Molecular Weight:
165.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

184-186 °C (lit.)

functional group

fluoro

SMILES string

OB(O)\C=C\c1cccc(F)c1

InChI

1S/C8H8BFO2/c10-8-3-1-2-7(6-8)4-5-9(11)12/h1-6,11-12H/b5-4+

InChI key

ONXKUGHKGCSZJO-SNAWJCMRSA-N

Application

Reactant for:
  • Enantioselective diastereoselective synthesis of tetracyclins via metal-catalyzed ring opening of meso-oxabicyclic alkenes followed by hydrogenation and Friedel-Crafts alkylation
  • Palladium-catalyzed Suzuki-Miyaura coupling reactions
  • Preparation of anticonvulsant agents
  • Preparation of piperazinyl pyrimidylhydroxamic acid deriatives as histone deacetylase inhibitors and antitumor agents
  • Preparation of ethyl-substituted conjugated dienoates and dienones by stereoselective Suzuki cross-coupling

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

637971-1G:
637971-BULK:
637971-VAR:
637971-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service