Skip to Content
Merck
All Photos(3)

Key Documents

Safety Information

634816

Sigma-Aldrich

4-(Methylthio)benzyl bromide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BrS
CAS Number:
Molecular Weight:
217.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

43-47 °C (lit.)

functional group

bromo
thioether

SMILES string

CSc1ccc(CBr)cc1

InChI

1S/C8H9BrS/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,6H2,1H3

InChI key

QDLKMRTVELBKFE-UHFFFAOYSA-N

Application

4-(Methylthio)benzyl bromide can undergo self-catalyzed oxidation in the presence of hydrogen peroxide to form the corresponding sulfoxide.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

634816-5G:
634816-BULK:
634816-VAR:
634816-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Self-Catalyzed Oxidation of Sulfides with Hydrogen Peroxide: A Green and Practical Process for the Synthesis of Sulfoxides"
Shi F, et al.
Advanced Synthesis & Catalysis, 349(16), 2425-2430 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service