Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

615617

Sigma-Aldrich

Sebacic acid-C-d16

98 atom % D, 99% (CP)

Synonym(s):

Decanedioic acid-C-d16

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CD2)8CO2H
Molecular Weight:
218.35
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

bp

294.5 °C (lit.)

mp

133-137 °C (lit.)

mass shift

M+16

SMILES string

[2H]C([2H])(C(O)=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C10H18O4/c11-9(12)7-5-3-1-2-4-6-8-10(13)14/h1-8H2,(H,11,12)(H,13,14)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2

InChI key

CXMXRPHRNRROMY-SADLKCKLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

615617-BULK:
615617-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service