615110
1-Pentanol-OD
≥98 atom % D, ≥99% (CP)
Synonym(s):
1-Pentan(ol-d), Amyl alcohol-OD, Pentyl alcohol-d
About This Item
Recommended Products
isotopic purity
≥98 atom % D
Assay
≥99% (CP)
bp
136-138 °C (lit.)
mp
-78 °C (lit.)
density
0.820 g/mL at 25 °C
mass shift
M+1
SMILES string
[2H]OCCCCC
InChI
1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3/i6D
InChI key
AMQJEAYHLZJPGS-RAMDWTOOSA-N
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
120.2 °F - closed cup
Flash Point(C)
49 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
FSL
Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
615110-BULK:
615110-VAR:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service