Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

604135

Sigma-Aldrich

Trichloroacetic-2-13C acid

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl313CCO2H
CAS Number:
Molecular Weight:
164.38
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

bp

196 °C (lit.)

mp

54-58 °C (lit.)

density

1.630 g/mL at 25 °C

mass shift

M+1

SMILES string

OC(=O)[13C](Cl)(Cl)Cl

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)/i2+1

InChI key

YNJBWRMUSHSURL-VQEHIDDOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

604135-VAR:
604135-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service