Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

598089

Sigma-Aldrich

Cyclopentadienyldicarbonyl(tetrahydrofuran)iron(II) tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13BF4FeO3
CAS Number:
Molecular Weight:
335.87
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: iron
reagent type: catalyst

mp

110 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Fe+].[C-]#[O+].[C-]#[O+].F[B-](F)(F)F.[CH]1[CH][CH][CH][CH]1.C2CCOC2

InChI

1S/C5H5.C4H8O.2CO.BF4.Fe/c2*1-2-4-5-3-1;2*1-2;2-1(3,4)5;/h1-5H;1-4H2;;;;/q;;;;-1;+1

InChI key

UEQAYXWNQGYVGN-UHFFFAOYSA-N

Application

Catalyst for:
  • Preparation of β-keto esters via Lewis acid-catalyzed addition of Et diazoacetate to arylaldehydes
  • Iron mediated nitrenoid transfer
  • Lewis acid catalyzed aziridination, cyclopropanation, and insertion reaction with diazo compounds
  • Diels-Alder reaction of dienes with enones

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

598089-BULK:
598089-VAR:
598089-250MG:4548173219141
598089-1G:4548173219158


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service