Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

596981

Sigma-Aldrich

Ethyl 1-amino-1-cyclobutanecarboxylate monohydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O2CC4H6NH2·HCl
CAS Number:
Molecular Weight:
179.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

SMILES string

Cl[H].CCOC(=O)C1(N)CCC1

InChI

1S/C7H13NO2.ClH/c1-2-10-6(9)7(8)4-3-5-7;/h2-5,8H2,1H3;1H

InChI key

YGAYWFFKRVMVBL-UHFFFAOYSA-N

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

596981-1G:
596981-BULK:
596981-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service