589640
Terephthaloyl-d4 chloride
98 atom % D
Synonym(s):
1,4-Benzene-2,3,5,6-d4-dicarbonyl dichloride
About This Item
Recommended Products
isotopic purity
98 atom % D
Quality Level
bp
266 °C
mp
79-81 °C (lit.)
mass shift
M+4
SMILES string
[2H]c1c([2H])c(c([2H])c([2H])c1C(Cl)=O)C(Cl)=O
InChI
1S/C8H4Cl2O2/c9-7(11)5-1-2-6(4-3-5)8(10)12/h1-4H/i1D,2D,3D,4D
InChI key
LXEJRKJRKIFVNY-RHQRLBAQSA-N
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 1
Flash Point(F)
356.0 °F - closed cup
Flash Point(C)
180 °C - closed cup
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PDSCL
Deleterious substance
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
589640-100MG:
589640-500MG:
589640-VAR:
589640-BULK:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service