Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

586986

Sigma-Aldrich

1-(2-Bromophenyl)-2-thiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7BrN2S
CAS Number:
Molecular Weight:
231.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

mp

125-129 °C (lit.)

SMILES string

NC(=S)Nc1ccccc1Br

InChI

1S/C7H7BrN2S/c8-5-3-1-2-4-6(5)10-7(9)11/h1-4H,(H3,9,10,11)

InChI key

QIGMVYSPXPXCPN-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

586986-5G:
586986-VAR:
586986-BULK:
586986-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service