Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

575763

Sigma-Aldrich

Sodium hydrogen periodate

99.999% trace metals basis

Synonym(s):

Sodium triparaperiodate, Trisodium dihydrogen orthoperiodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3H2IO6
CAS Number:
Molecular Weight:
293.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: oxidant

SMILES string

[Na+].[Na+].[Na+].OI(O)([O-])([O-])([O-])=O

InChI

1S/H5IO6.3Na/c2-1(3,4,5,6)7;;;/h(H5,2,3,4,5,6,7);;;/q;3*+1/p-3

InChI key

DSYMXPNUZLZKOA-UHFFFAOYSA-K

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 1: Oxidizing solids
Periodates
Hazardous rank I
1st oxidizing solid

JAN Code

575763-20G:4548173995083
575763-50G:4548173995090
575763-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service