Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

565105

Sigma-Aldrich

N-(1-Naphthyl)-N-phenylacrylamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15NO
CAS Number:
Molecular Weight:
273.33
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

contains

~1000 ppm MEHQ as additive

mp

102-113 °C (lit.)

λmax

221 nm

storage temp.

2-8°C

SMILES string

C=CC(=O)N(c1ccccc1)c2cccc3ccccc23

InChI

1S/C19H15NO/c1-2-19(21)20(16-11-4-3-5-12-16)18-14-8-10-15-9-6-7-13-17(15)18/h2-14H,1H2

InChI key

ZAAYATMUFGQSPN-UHFFFAOYSA-N

Application

Fluorescent monomer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

565105-VAR:
565105-100MG:
565105-BULK:
565105-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service