Skip to Content
Merck
All Photos(1)

Documents

Safety Information

554405

Sigma-Aldrich

3-(Difluoromethoxy)benzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2CHOC6H4SO2Cl
CAS Number:
Molecular Weight:
242.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5100 (lit.)

bp

233-234 °C (lit.)

density

1.509 g/mL at 25 °C (lit.)

SMILES string

FC(F)Oc1cccc(c1)S(Cl)(=O)=O

InChI

1S/C7H5ClF2O3S/c8-14(11,12)6-3-1-2-5(4-6)13-7(9)10/h1-4,7H

InChI key

VKSMJFRQQMGYHS-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

208.0 °F - closed cup

Flash Point(C)

97.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

554405-BULK:
554405-5G:
554405-1G:
554405-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service