Recommended Products
Assay
97%
refractive index
n20/D 1.502 (lit.)
bp
196-197 °C (lit.)
density
1.585 g/mL at 25 °C (lit.)
SMILES string
FC(F)Oc1cccc(Br)c1
InChI
1S/C7H5BrF2O/c8-5-2-1-3-6(4-5)11-7(9)10/h1-4,7H
InChI key
NWBDGKNKAFGQQL-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
553433-1G:
553433-BULK:
553433-VAR:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service