Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

552607

Sigma-Aldrich

3-(1-Piperazino)propyl functionalized silica gel

200-400 mesh, extent of labeling: 0.8 mmol/g loading

Synonym(s):

Piperazino-3 functionalized silica gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.22

extent of labeling

0.8 mmol/g loading

surface area

500 m2/g

particle size

200-400 mesh

pore size

60 Å pore size

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

552607-100G:
552607-25G:
552607-10G:
552607-VAR:
552607-BULK:
552607-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Functionalized silica gel is used in size-exclusion chromatography of cationic polymers and in the separation of sugars, steroids, cholesterol, and triglycerides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service