Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

548057

Sigma-Aldrich

2-Fluorobiphenyl-4-carboxaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H3(F)COH
CAS Number:
Molecular Weight:
200.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

27-31 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1cc(C=O)ccc1-c2ccccc2

InChI

1S/C13H9FO/c14-13-8-10(9-15)6-7-12(13)11-4-2-1-3-5-11/h1-9H

InChI key

BBFKVFLWRKZSPM-UHFFFAOYSA-N

General description

2-Fluorobiphenyl-4-carboxaldehyde is an oxygen-containing polycyclic aromatic hydrocarbon derivative (OPAH).

Application

2-Fluorobiphenyl-4-carboxaldehyde may be used as a starting material in the synthesis of (2-fluoro-1,1′-biphenyl-4-yl)acetic acid, a novel unsubstituted flurbiprofen (FLU) derivative.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

548057-1G:
548057-5G:
548057-VAR:
548057-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piera Sozio et al.
Molecules (Basel, Switzerland), 18(9), 10747-10767 (2013-09-06)
Alzheimer's disease (AD) is characterized by irreversible and progressive loss of memory and cognition and profound neuronal loss. Current therapeutic strategies for the treatment of AD have been directed to a variety of targets with the aim of reversing or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service