Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

546917

Sigma-Aldrich

2,3-Dichlorobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3SO2Cl
CAS Number:
Molecular Weight:
245.51
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

60-64 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

Clc1cccc(c1Cl)S(Cl)(=O)=O

InChI

1S/C6H3Cl3O2S/c7-4-2-1-3-5(6(4)8)12(9,10)11/h1-3H

InChI key

PQODWTNHDKDHIW-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 2 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

49.0 °F - closed cup

Flash Point(C)

9.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 2: Flammable solids
Flammable solid
Hazardous rank III

JAN Code

546917-5G:
546917-VAR:
546917-BULK:
546917-25G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service