Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

537519

Sigma-Aldrich

2-Butoxyethyl acetate

99%

Synonym(s):

1-Acetoxy-2-butoxyethane, BCA, Ethylene glycol monobutyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
160.21
Beilstein:
1756960
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

vapor density

5.5 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)

Assay

99%

autoignition temp.

644 °F

expl. lim.

0.88 %, 33 °F
8.54 %, 135 °F

impurities

≤0.050% water
water

refractive index

n20/D 1.413 (lit.)

bp

192 °C (lit.)

mp

-63 °C (lit.)

density

0.942 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCOC(C)=O

InChI

1S/C8H16O3/c1-3-4-5-10-6-7-11-8(2)9/h3-7H2,1-2H3

InChI key

NQBXSWAWVZHKBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water soluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

537519-18L:
537519-VAR:
537519-20L:
537519-1L:
537519-4L:
537519-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service