Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

536113

Sigma-Aldrich

2-Chlorotrityl amine, polymer-bound

100-200 mesh, extent of labeling: 1.3-1.8 mmol/g loading, 1 % cross-linked

Synonym(s):

2-Chlorotriphenylmethyl amine resin, Amino-2-chlorotrityl resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

crosslinking

1 % cross-linked

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

1.3-1.8 mmol/g loading

particle size

100-200 mesh

functional group

amine

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.NC(c3ccccc3)(c4ccccc4)c5ccccc5Cl

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

536113-BULK:
536113-5G:
536113-1G:
536113-25G:
536113-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service