Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

528676

Sigma-Aldrich

Ethyl 2,4-diethoxybenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2C6H3CO2C2H5
CAS Number:
Molecular Weight:
238.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5195 (lit.)

bp

120 °C (lit.)

density

1.059 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1ccc(OCC)cc1OCC

InChI

1S/C13H18O4/c1-4-15-10-7-8-11(13(14)17-6-3)12(9-10)16-5-2/h7-9H,4-6H2,1-3H3

InChI key

XCKIALVAGKRVNP-UHFFFAOYSA-N

General description

Ethyl 2,4-diethoxybenzoate is an ethyl ester of 2,4-diethoxybenzoic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

528676-BULK:
528676-5G:
528676-VAR:
528676-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service