Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

517976

Sigma-Aldrich

Amberjet® 4200 chloride form

chloride form, beads

Synonym(s):

Poly(styrene-co-divinylbenzene) with quaternary amine functionality, chloride form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

beads

crosslinking

1 % cross-linked

specific activity

3.5-4.5 mmol Cl-/g

parameter

60 °C Cl- form max. temp.

impurities

50-60%

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium functional group

particle size

600-800 μm

capacity

≥1.30 meq/mL by wetted bed volume

SMILES string

[Cl-].C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C[N+](C)(C)c3ccc(C=C)cc3

Looking for similar products? Visit Product Comparison Guide

Application

Ion exchange resin (strongly basic anion exchanger)
Strongly basic (type I) anion exchange resin for water demineralization; useful for removal of iron and zinc from hydrochloric acid, and recovery of silver from photographic effluents.

Legal Information

Amberjet is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

517976-BULK:
517976-100G:
517976-500G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service