515019
5-Bromo-2-methylaniline
97%
Synonym(s):
5-Bromo-o-toluidine
About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.614 (lit.)
bp
139 °C/17 mmHg (lit.)
mp
33 °C (lit.)
density
1.49 g/mL at 25 °C (lit.)
SMILES string
Cc1ccc(Br)cc1N
InChI
1S/C7H8BrN/c1-5-2-3-6(8)4-7(5)9/h2-4H,9H2,1H3
InChI key
RXQNKKRGJJRMKD-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
515019-BULK:
515019-VAR:
515019-25G:
515019-5G:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service