Skip to Content
Merck
All Photos(2)

Documents

Safety Information

512869

Sigma-Aldrich

3-Formyl-4-methoxyphenylboronic acid

≥95%

Synonym(s):

3-Formyl-4-methoxybenzeneboronic acid, 5-Dihydroxyboryl-2-methoxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOC6H3(OCH3)B(OH)2
CAS Number:
Molecular Weight:
179.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

mp

214-217 °C (lit.)

SMILES string

COc1ccc(cc1C=O)B(O)O

InChI

1S/C8H9BO4/c1-13-8-3-2-7(9(11)12)4-6(8)5-10/h2-5,11-12H,1H3

InChI key

YJQDBKGGRPJSOI-UHFFFAOYSA-N

Application

Reactant for:
  • Copper(I)-mediated cyanation
  • Suzuki-Miyaura cross-coupling reactions
  • Preparation of biologically and pharmacologically active molecules

Other Notes

Contains varying amounts of anhydride

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

JAN Code

512869-25G:
512869-BULK:
512869-5G:
512869-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service