Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

511234

Sigma-Aldrich

Cyclopentadienyltungsten(II) tricarbonyl hydride

98%

Synonym(s):

Tricarbonyl(cyclopentadienyl)hydridotungsten(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O3W
CAS Number:
Molecular Weight:
333.97
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98%

reaction suitability

core: tungsten
reagent type: catalyst

storage temp.

2-8°C

SMILES string

[WH].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1

InChI

1S/C5H5.3CO.W.H/c1-2-4-5-3-1;3*1-2;;/h1-5H;;;;;

InChI key

FYSFWBGCVRRUIX-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1 - Water-react 3

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

170.0 °F - closed cup

Flash Point(C)

76.67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

511234-BULK:
511234-VAR:
511234-1G:
511234-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service