Skip to Content
Merck
All Photos(1)

Key Documents

511226

Sigma-Aldrich

Cyclopentadienylmolybdenum(I) tricarbonyl hydride

Synonym(s):

Tricarbonyl(cyclopentadienyl)hydridomolybdenum(I)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6MoO3
CAS Number:
Molecular Weight:
246.07
UNSPSC Code:
12352300
PubChem Substance ID:

SMILES string

[MoH].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service