Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

510327

Sigma-Aldrich

Trichloroacetimidate on Wang resin

100-200 mesh, extent of labeling: 0.75-1.25 mmol/g trichloroacetimidate loading, 1 % cross-linked

Synonym(s):

4-Alkoxybenzyl trichloroacetimidate, polymer-bound, Wang trichloroacetimidate resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

crosslinking

1 % cross-linked

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

0.75-1.25 mmol/g trichloroacetimidate loading

particle size

100-200 mesh

storage temp.

2-8°C

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.ClC(Cl)(Cl)C(=N)OCc3ccc(OCc4ccccc4)cc3

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

510327-1G:
510327-10G:
510327-50G:
510327-BULK:
510327-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service