Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

497398

Sigma-Aldrich

Calcium isopropoxide

powder, ≥99% trace metals basis

Synonym(s):

Calcium isopropylate, Diisopropoxy calcium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OCH(CH3)2)2
CAS Number:
Molecular Weight:
158.25
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99% trace metals basis

form

powder

reaction suitability

core: calcium

SMILES string

CC(C)O[Ca]OC(C)C

InChI

1S/2C3H7O.Ca/c2*1-3(2)4;/h2*3H,1-2H3;/q2*-1;+2

InChI key

MMLSWLZTJDJYJH-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Self-heat. 2 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

497398-BULK:
497398-10G:
497398-VAR:
497398-10G-PW:
497398-2G:
497398-2G-PW:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service