Skip to Content
Merck
All Photos(1)

Key Documents

496685

Sigma-Aldrich

2,2,6,6-Tetramethyl-1-(1-phenylethoxy)-4-piperidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO2
CAS Number:
Molecular Weight:
277.40
MDL number:
UNSPSC Code:
12162002

SMILES string

CC(ON1C(C)(C)CC(O)CC1(C)C)c2ccccc2

Application

Initiator for stable free radical polymerizations

Features and Benefits

For accurate control of MW and polydispersity.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macromolecules, 30, 1929-1929 (1997)
Macromolecules, 26, 2987-2987 (1993)
Macromolecules, 29, 5245-5245 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service