Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

495999

Sigma-Aldrich

Tris(butylcyclopentadienyl)erbium(III)

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Er(C5H4C4H9)3
CAS Number:
Molecular Weight:
530.86
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.9% trace metals basis

form

liquid

reaction suitability

core: erbium
reagent type: catalyst

bp

240 °C/0.1 mmHg (lit.)

density

1.309 g/mL at 25 °C (lit.)

SMILES string

[Er].CCCC[C]1[CH][CH][CH][CH]1.CCCC[C]2[CH][CH][CH][CH]2.CCCC[C]3[CH][CH][CH][CH]3

InChI

1S/3C9H13.Er/c3*1-2-3-6-9-7-4-5-8-9;/h3*4-5,7-8H,2-3,6H2,1H3;

InChI key

JRCMYZNYPKTOPG-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Pyr. Liq. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 3: Spontaneously combustible substances and water- reactive materials
Materials containing Organometallic compounds
Hazardous rank I
1st spontaneously combustible materials and water reactive materials

JAN Code

495999-5G:
495999-BULK:
495999-VAR:
495999-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service