Skip to Content
Merck
All Photos(1)

Documents

Safety Information

495018

Sigma-Aldrich

1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

159-162 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CCCC(C1)C(O)=O

InChI

1S/C11H19NO4/c1-11(2,3)16-10(15)12-6-4-5-8(7-12)9(13)14/h8H,4-7H2,1-3H3,(H,13,14)

InChI key

NXILIHONWRXHFA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

495018-BULK:
495018-VAR:
495018-25G:
495018-5G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bunta Watanabe et al.
PloS one, 10(8), e0136242-e0136242 (2015-08-22)
CCG-1423 suppresses several pathological processes including cancer cell migration, tissue fibrosis, and the development of atherosclerotic lesions. These suppressions are caused by inhibition of myocardin-related transcription factor A (MRTF-A), which is a critical factor for epithelial-mesenchymal transition (EMT). CCG-1423 can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service