Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

483400

Sigma-Aldrich

4-Butylphenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C6H4NCO
CAS Number:
Molecular Weight:
175.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.517 (lit.)

bp

108 °C/3 mmHg (lit.)

density

0.992 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCc1ccc(cc1)N=C=O

InChI

1S/C11H13NO/c1-2-3-4-10-5-7-11(8-6-10)12-9-13/h5-8H,2-4H2,1H3

InChI key

LJJRXPXDTAUVQU-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

483400-VAR:
483400-25ML:
483400-BULK:
483400-5ML:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glenn E Schafe et al.
The European journal of neuroscience, 22(1), 201-211 (2005-07-21)
Nitric oxide (NO) has been widely implicated in synaptic plasticity and memory formation. In studies of long-term potentiation (LTP), NO is thought to serve as a 'retrograde messenger' that contributes to presynaptic aspects of LTP expression. In this study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service