Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

47809

Sigma-Aldrich

Fmoc-Phe(2,4-Cl2)-OH

≥98.0% (HPLC)

Synonym(s):

Fmoc-2,4-dichloro-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H19Cl2NO4
CAS Number:
Molecular Weight:
456.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (HPLC)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccc(Cl)cc1Cl)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C24H19Cl2NO4/c25-15-10-9-14(21(26)12-15)11-22(23(28)29)27-24(30)31-13-20-18-7-3-1-5-16(18)17-6-2-4-8-19(17)20/h1-10,12,20,22H,11,13H2,(H,27,30)(H,28,29)/t22-/m0/s1

InChI key

PGIMVGROPOKRNA-QFIPXVFZSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

47809-1G:
47809-5G:
47809-BULK:
47809-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service