Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

47719

Sigma-Aldrich

N-Formylglycine ethyl ester

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONHCH2COOC2H5
CAS Number:
Molecular Weight:
131.13
Beilstein:
1099378
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.453 (lit.)
n20/D 1.453

bp

267-269 °C (lit.)

density

1.15 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCOC(=O)CNC=O

InChI

1S/C5H9NO3/c1-2-9-5(8)3-6-4-7/h4H,2-3H2,1H3,(H,6,7)

InChI key

GMBCCEOJUWMBPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

47719-INTR:
47719-50ML:
47719-10ML:
47719-VAR:
47719-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service