Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

47554

Sigma-Aldrich

1-Fmoc-4-piperidineacetic acid

≥99.0% (HPLC)

Synonym(s):

1-Fmoc-piperidin-4-ylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23NO4
CAS Number:
Molecular Weight:
365.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥99.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

135-138 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)CC1CCN(CC1)C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C22H23NO4/c24-21(25)13-15-9-11-23(12-10-15)22(26)27-14-20-18-7-3-1-5-16(18)17-6-2-4-8-19(17)20/h1-8,15,20H,9-14H2,(H,24,25)

InChI key

CQAGUAUKAZHOSU-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

47554-1G-F:
47554-BULK-F:
47554-250MG-F:
47554-VAR-F:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service