Skip to Content
Merck
All Photos(1)

Key Documents

468169

Sigma-Aldrich

N-Ethyl-N-[(heptadecafluorooctyl)sulfonyl]glycine potassium salt solution

42 wt. % in water: 2-butoxyethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7SO2N(C2H5)CH2CO2K
CAS Number:
Molecular Weight:
623.32
MDL number:
UNSPSC Code:
12352300

contains

~30% water

composition

2-butoxyethanol and C-4 to C-7 fluoroalkyl groups., 14%

concentration

42 wt. % in water: 2-butoxyethanol

refractive index

n20/D 1.371

pH

8-11

viscosity

30 cP(25 °C)(lit.)

bp

100 °C

density

1.3 g/mL at 25 °C

SMILES string

[K+].CCN(CC([O-])=O)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Application

Anionic fluorosurfactant.
Leveling and wetting agent where unusual chemical stability is required.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service