467529
1H-Benzotriazole-1-acetonitrile
95%
Synonym(s):
Benzotriazol-1-ylacetonitrile
About This Item
Assay
95%
reaction suitability
reaction type: C-C Bond Formation
mp
84-89 °C (lit.)
SMILES string
N#CCn1nnc2ccccc12
InChI
1S/C8H6N4/c9-5-6-12-8-4-2-1-3-7(8)10-11-12/h1-4H,6H2
InChI key
DIFOLZGBHRDEFT-UHFFFAOYSA-N
Application
- Hydrothermal reactions
- Preparation of aminonaphthylacrylonitrile derivatives as green luminance dyes
- Preparation of (aminomethyl)(cyanovinylphenyl)oxazolidinones with heterocyclic pendant as antibacterial agents
- Cyclization reactions with sodium azide
- Preparation of substituted triazoles (as antitumor agents) via rearrangement reaction followed by cycloaddition of corresponding (cyanomethyl)benzotriazole
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PDSCL
Deleterious substance
JAN Code
467529-250MG:
467529-VAR:
467529-1G:4548173155449
467529-5G:4548173155456
467529-BULK:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service