Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

456411

Sigma-Aldrich

Titanium(III) chloride-aluminum chloride

TiCl3:AlCl3 3:1

Synonym(s):

Aluminum titanium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(TiCl3)3 · AlCl3
CAS Number:
Molecular Weight:
596.02
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

solid

composition

TiCl3:AlCl3, 3:1

reaction suitability

reagent type: catalyst
core: titanium

SMILES string

Cl[Al](Cl)Cl.Cl[Ti](Cl)Cl.Cl[Ti](Cl)Cl.Cl[Ti](Cl)Cl

InChI

1S/Al.12ClH.3Ti/h;12*1H;;;/q+3;;;;;;;;;;;;;3*+3/p-12

InChI key

DGFMFXMKCUOBCD-UHFFFAOYSA-B

Application

Low-valent titanium is a useful reducing agent in the synthesis of imidazolines.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Pyr. Sol. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

456411-BULK:
456411-250G:
456411-VAR:
456411-50G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atul A More et al.
Organic letters, 18(6), 1458-1461 (2016-03-08)
The first total synthesis of integrastatin B, a potent HIV-1 integrase inhibitor, has been accomplished in seven steps with a 17.9% overall yield employing easily accessible starting compounds. The Oxone-mediated oxidative benzofuran dearomatization cascade has been employed as the key
Periasamy, M. et al.
Tetrahedron Letters, 37, 4767-4767 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service