Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

454168

Sigma-Aldrich

4-Fluoro-4′-(phenylethynyl)benzophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CC6H4COC6H4F
CAS Number:
Molecular Weight:
300.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

151-153 °C (lit.)

SMILES string

Fc1ccc(cc1)C(=O)c2ccc(cc2)C#Cc3ccccc3

InChI

1S/C21H13FO/c22-20-14-12-19(13-15-20)21(23)18-10-8-17(9-11-18)7-6-16-4-2-1-3-5-16/h1-5,8-15H

InChI key

SKWWXACONTVUHM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

454168-5G:
454168-VAR:
454168-1G:
454168-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endgroup substituent effects on the rate/extent of network formation and adhesion for phenylethynyl-terminated poly (arylene ether sulfone) oligomers.
Ayambem A, et al.
Polymer, 41(13), 5109-5124 (2000)
Synthesis and Characterization of Hyperbranched Poly (Arylene-Ether-Ketone-Imide)s Containing Phenylethynyl Groups.
Wang DH, et al.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 46(1), 793-794 (2005)
New hexaarylbenzene-containing di-and tetrafluoroarenes.
Keshtov ML, et al.
Russian Chemical Bulletin, 54(8), 1939-1941 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service