Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

452661

Sigma-Aldrich

N-(Benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine

97%

Synonym(s):

Benzyl N-[imino(1-pyrazolyl)methyl]carbamate, Z-1H-pyrazole-1-carboxamidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N4O2
CAS Number:
Molecular Weight:
244.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

106-110 °C (lit.)

SMILES string

N=C(NC(=O)OCc1ccccc1)n2cccn2

InChI

1S/C12H12N4O2/c13-11(16-8-4-7-14-16)15-12(17)18-9-10-5-2-1-3-6-10/h1-8H,9H2,(H2,13,15,17)

InChI key

BXUMISWXFKCEIA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

452661-10G:
452661-VAR:
452661-1G:
452661-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service